(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 1-(tert-Butoxycarbonyl)-L-pipecolic AcidBoc-HomoPro-OHBoc-Pip-OH1-Boc-L-pipecolic Acid(S)-N-BOC-Piperidine-2-carboxylic acid(S)-1-Boc-2-piperidinecarboxylic AcidBoc-Homopro-OH

APOSOR14684-5GEA 0 EUR
APOSOR14684-5G APOSOR14684-1G
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
Formula: C₁₁H₁₉NO₄
MW: 229,28 g/mol
Melting Pt: 121…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00151904
CAS Number: 26250-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR