3-(2-Naphthyl)-L-alanine

Warning

Synonyms: (S)-alpha-Amino-2-naphthalenepropionic Acid(S)-2-Amino-3-(naphthalen-2-yl)propanoic acid

APOSOR14713-1GEA 0 EUR
APOSOR14713-1G APOSOR14713-25G APOSOR14713-5G
3-(2-Naphthyl)-L-alanine
3-(2-Naphthyl)-L-alanine
Formula: C₁₃H₁₃NO₂
MW: 215,25 g/mol
Melting Pt: 240 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00066087
CAS Number: 58438-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR