2-Mercapto-1,3,4-thiadiazole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1,3,4-Thiadiazole-2-thiol
APOSOR15040-5GEA
14.7
EUR
APOSOR15040-5G
2-Mercapto-1,3,4-thiadiazole
2-Mercapto-1,3,4-thiadiazole
Formula:
C₂H₂N₂S₂ MW: 118,17 g/mol Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040404 CAS Number: 18686-82-3 EINECS: 242-504-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...