5-Amino-3-(3-bromophenyl)-1H-pyrazole

Supplier: Apollo Scientific
Danger

OR15137-1G
APOSOR15137-1GEA 82.9 EUR
APOSOR15137-1G
5-Amino-3-(3-bromophenyl)-1H-pyrazole
5-Amino-3-(3-bromophenyl)-1H-pyrazole
Formula: C₉H₈BrN₃
Melting Pt: 149…151 °C
Storage Temperature: Ambient
MDL Number: MFCD06738756
CAS Number: 149246-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR