3-(3,5-Dimethoxyphenyl)-1H-pyrazol-5-amine

Supplier: Apollo Scientific

Synonyms: 5-Amino-3-(3,5-dimethoxyphenyl)-1H-pyrazole

OR15177-1G OR15177-5G
APOSOR15177-1GEA 205 EUR
APOSOR15177-1G APOSOR15177-5G
3-(3,5-Dimethoxyphenyl)-1H-pyrazol-5-amine
3-(3,5-Dimethoxyphenyl)-1H-pyrazol-5-amine
Formula: C₁₁H₁₃N₃O₂
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD08443954
CAS Number: 569655-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR