2-Hydrazino-6-nitrobenzothiazole

APOSOR15203-1GEA 0 EUR
APOSOR15203-1G
2-Hydrazino-6-nitrobenzothiazole
2-Hydrazino-6-nitrobenzothiazole
Formula: C₇H₆N₄O₂S
Melting Pt: 273…274 °C
Storage Temperature: Ambient
MDL Number: MFCD00022864
CAS Number: 30710-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR