2-Ethoxy-4-(tributylstannyl)thiazole
APOSOR15534-500MGEA
268
EUR
APOSOR15534-500MG
2-Ethoxy-4-(tributylstannyl)thiazole
2-Ethoxy-4-(tributylstannyl)thiazole
Formula:
C₁₇H₃₃NOSSn MW: 418,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07787373 CAS Number: 240816-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...