4-(Tributylstannyl)thiazole 95%
|
Danger
|
APOSOR15546-250MGEA
143
EUR
APOSOR15546-250MG
APOSOR15546-5G
APOSOR15546-1G
4-(Tributylstannyl)thiazole 95%
4-(Tributylstannyl)thiazole
Formula:
C₁₅H₂₉NSSn MW: 374,17 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01319073 CAS Number: 173979-01-6 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...