cis-1-Ethoxy-2-(tributylstannyl)ethene

Supplier: Apollo Scientific
Danger

APOSOR15604-250MGEA 42.6 EUR
APOSOR15604-250MG
cis-1-Ethoxy-2-(tributylstannyl)ethene
cis-1-Ethoxy-2-(tributylstannyl)ethene
Formula: C₁₆H₃₄OSn
MW: 361,15
Boiling Pt: 84…87 °C/1mmHg
Density: 1,082 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07787391
CAS Number: 64724-29-4
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR