6-(Benzyloxy)-1H-indole 98%
Supplier: Apollo Scientific
|
Warning
|
APOSOR1675-1GEA
14.7
EUR
APOSOR1675-1G
6-(Benzyloxy)-1H-indole 98%
6-(Benzyloxy)-1H-indole
Formula:
C15H13NO MW: 223,28 g/mol Melting Pt: 114…117 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00053554 CAS Number: 15903-94-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...