tert-Butyl-2-((diphenylmethylene)amino)acetate 98%

Supplier: Apollo Scientific

Synonyms: Glycine, N-(diphenylmethylene)-, 1,1-dimethylethyl esterN-(Diphenylmethylene)glycine tert-butyl ester

APOSOR17705-100GEA 139 EUR
APOSOR17705-100G
tert-Butyl-2-((diphenylmethylene)amino)acetate 98%
tert-Butyl-2-((diphenylmethylene)amino)acetate
Formula: C₁₉H₂₁NO₂
MW: 295,382 g/mol
Melting Pt: 114…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00134280
CAS Number: 81477-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR