(2-Carboxyethyl)triphenylphosphonium bromide 97%

Supplier: Apollo Scientific
Warning

APOSOR18825-25GEA 88.9 EUR
APOSOR18825-25G
(2-Carboxyethyl)triphenylphosphonium bromide 97%
(2-Carboxyethyl)triphenylphosphonium bromide
Formula: C₂₁H₂₀BrO₂P
Melting Pt: 198 °C
Flash Pt: >230 °C
Storage Temperature: Ambient
MDL Number: MFCD00031698
CAS Number: 51114-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR