3-Quinolineboronic acid hydrochloride 98%
|
Warning
|
APOSOR1932-100GEA
0
EUR
APOSOR1932-100G
APOSOR1932-5G
APOSOR1932-25G
3-Quinolineboronic acid hydrochloride 98%
3-Quinolineboronic acid hydrochloride
Formula:
C₉H₉BClNO₂ MW: 209,44 g/mol Melting Pt: 190…194 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659836 CAS Number: 850568-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...