Ethyl-2,4-dimethylthiazole-5-carboxylate
Supplier: Apollo Scientific
Synonyms:
Ethyl 2,4-dimethylthiazole-5-carboxylate
APOSOR200011-10GEA
136
EUR
APOSOR200011-10G
APOSOR200011-1G
Ethyl-2,4-dimethylthiazole-5-carboxylate
Ethyl 2,4-dimethylthiazole-5-carboxylate
Formula:
C₈H₁₁NO₂S MW: 185,24 g/mol Melting Pt: 46…50 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052874 CAS Number: 7210-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...