5-Chlorothiophene-2-carbonyl chloride 97%
Supplier: Apollo Scientific
|
Danger
|
APOSOR200094-25GEA
87.3
EUR
APOSOR200094-25G
5-Chlorothiophene-2-carbonyl chloride 97%
5-Chlorothiophene-2-carbonyl chloride
Formula:
C5H2Cl2OS MW: 181,04 g/mol Melting Pt: 4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043886 CAS Number: 42518-98-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...