1-Bromo-4-hexyloxybenzene

Supplier: Apollo Scientific

Synonyms: 1-Bromo-4-(hexyloxy)benzene1-(4-Bromophenoxy)hexane

APOSOR21905-50GEA 285 EUR
APOSOR21905-50G APOSOR21905-10G
1-Bromo-4-hexyloxybenzene
1-Bromo-4-hexyloxybenzene
Formula: C₁₂H₁₇BrO
MW: 257,18 g/mol
Boiling Pt: 110 °C/0,1mmHg
Density: 1,23 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00173753
CAS Number: 30752-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR