2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone
|
Danger
|
Synonyms:
3,5-Bis(tert-butyl)-4-hydroxyphenacyl bromide
, 2-Bromo-1-[3,5-di(tert-butyl)-4-hydroxyphenyl]ethan-1-one
APOSOR21940-10GEA
0
EUR
APOSOR21940-10G
2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone
2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone
Formula:
C₁₆H₂₃BrO₂ MW: 327,26 g/mol Melting Pt: 104…107 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00173889 CAS Number: 14386-64-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...