Methyl-5-acetylsalicylate

Synonyms: Methyl 5-acetyl-2-hydroxybenzoate

APOSOR22086-100GEA 0 EUR
APOSOR22086-100G APOSOR22086-25G APOSOR22086-5G
Methyl-5-acetylsalicylate
Methyl 5-acetylsalicylate
Formula: C₁₀H₁₀O₄
MW: 194,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040924
CAS Number: 16475-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR