((3-Bromopropoxy)methyl)benzene 96%

Warning

Synonyms: Benzyl 3-bromopropyl ether3-bromopropylbenzyl ether

APOSOR22888-10GEA 82.9 EUR
APOSOR22888-10G APOSOR22888-25G APOSOR22888-100G
((3-Bromopropoxy)methyl)benzene 96%
((3-Bromopropoxy)methyl)benzene
Formula: C₁₀H₁₃BrO
MW: 229,117 g/mol
Boiling Pt: 130…132 °C/8mmHg
Density: 1,298 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00134570
CAS Number: 54314-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR