(3,5-Dimethyl-4-isoxazolyl)methanol

Supplier: Apollo Scientific
Warning

Synonyms: 3,5-Dimethyl-4-(hydroxymethyl)isoxazole

APOSOR22989-1GEA 121 EUR
APOSOR22989-1G APOSOR22989-5G
(3,5-Dimethyl-4-isoxazolyl)methanol
(3,5-Dimethyl-4-isoxazolyl)methanol
Formula: C₆H₉NO₂
Melting Pt: 76…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00464226
CAS Number: 19788-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR