6-Bromoquinoxaline

Supplier: Apollo Scientific
Danger

APOSOR23162-1GEA 68.9 EUR
APOSOR23162-1G APOSOR23162-500MG APOSOR23162-5G
6-Bromoquinoxaline
6-Bromoquinoxaline
Formula: C₈H₅BrN₂
Melting Pt: 48…53 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00837757
CAS Number: 50998-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR