3-Thiophenesulphonyl chloride

Supplier: Apollo Scientific
Danger

APOSOR23200-1GEA 52.6 EUR
APOSOR23200-1G
3-Thiophenesulphonyl chloride
3-Thiophenesulphonyl chloride
Formula: C₄H₃ClO₂S₂
Boiling Pt: 101…103 °C
Melting Pt: 44…46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02682011
CAS Number: 51175-71-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR