6-Methoxy-8-nitroquinoline 98%
|
Warning
|
APOSOR2347-25GEA
0
EUR
APOSOR2347-25G
APOSOR2347-10G
APOSOR2347-100G
6-Methoxy-8-nitroquinoline 98%
6-Methoxy-8-nitroquinoline
Formula:
C₁₀H₈N₂O₃ MW: 204,1800 g/mol Melting Pt: 158…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006802 CAS Number: 85-81-4 EINECS: 201-633-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...