2-Furoylacetonitrile

Supplier: Apollo Scientific
Warning

Synonyms: 3-(Fur-2-yl)-3-oxopropanenitrile

APOSOR23643-1GEA 25.1 EUR
APOSOR23643-1G APOSOR23643-5G
2-Furoylacetonitrile
2-Furoylacetonitrile
Formula: C₇H₅NO₂
MW: 135,12
Melting Pt: 77…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00052210
CAS Number: 31909-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR