1,4-Dioxane-2,3-diol

Warning

Synonyms: 2,3-Dihydroxy-1,4-dioxane

APOSOR23815-25GEA 0 EUR
APOSOR23815-25G APOSOR23815-100G APOSOR23815-5G
1,4-Dioxane-2,3-diol
1,4-Dioxane-2,3-diol
Formula: C₄H₈O₄
MW: 120,10 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00006572
CAS Number: 4845-50-5
EINECS: 225-431-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR