Methyl quinoxaline-6-carboxylate
Supplier: Apollo Scientific
Synonyms:
Methyl 6-Quinoxalinecarboxylate
APOSOR26804-10GEA
93.1
EUR
APOSOR26804-10G
Methyl quinoxaline-6-carboxylate
Methyl quinoxaline-6-carboxylate
Formula:
C₁₀H₈N₂O₂ MW: 188,19 g/mol Melting Pt: 101…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00102618 CAS Number: 23088-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...