5-Methyl-2,1,3-benzothiadiazole
Supplier: Apollo Scientific
|
Warning
|
APOSOR26809-250MGEA
14.7
EUR
APOSOR26809-250MG
5-Methyl-2,1,3-benzothiadiazole
5-Methyl-2,1,3-benzothiadiazole
Formula:
C₇H₆N₂S Boiling Pt: 62…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173945 CAS Number: 1457-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...