2-Bromo-2'-chloroacetanilide

Supplier: Apollo Scientific
Warning

Synonyms: N-(Bromoacetyl)-2-chloroaniline

APOSOR2782-1GEA 36.4 EUR
APOSOR2782-1G APOSOR2782-5G
2-Bromo-2'-chloroacetanilide
2-Bromo-2'-chloroacetanilide
Formula: C₈H₇BrClNO
Melting Pt: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00173815
CAS Number: 5439-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR