2'-Benzoyl-2,4'-dichloroacetanilide

Supplier: Apollo Scientific

Synonyms: N1-(2-Benzoyl-4-chlorophenyl)-2-chloroacetamide

APOSOR28428-25GEA 626 EUR
APOSOR28428-25G APOSOR28428-5G
2'-Benzoyl-2,4'-dichloroacetanilide
2'-Benzoyl-2,4'-dichloroacetanilide
Formula: C₁₅H₁₁Cl₂NO₂
Melting Pt: 119…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00018905
CAS Number: 4016-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR