2-Chloro-4-methoxybenzonitrile 99%

Supplier: Apollo Scientific
Warning

APOSOR29211-5GEA 17.6 EUR
APOSOR29211-5G APOSOR29211-1G APOSOR29211-25G
2-Chloro-4-methoxybenzonitrile 99%
2-Chloro-4-methoxybenzonitrile
Formula: C₈H₆ClNO
MW: 167,5900 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00156133
CAS Number: 127666-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR