4'-(4-Bromophenyl)acetophenone 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Acetyl-4'-bromobiphenyl
, 1-(4'-Bromo-[1,1'-biphenyl]-4-yl)ethanone
APOSOR2964-25GEA
115
EUR
APOSOR2964-25G
4'-(4-Bromophenyl)acetophenone 98%
4'-(4-Bromophenyl)acetophenone
Formula:
C₁₄H₁₁BrO MW: 275,15 g/mol Melting Pt: 129…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143242 CAS Number: 5731-01-1 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...