3-Bromobenzaldehyde ethylene acetal 97%
Synonyms:
2-(3-Bromophenyl)-1,3-dioxolane
APOSOR30032-5GEA
0
EUR
APOSOR30032-5G
APOSOR30032-25G
APOSOR30032-100G
3-Bromobenzaldehyde ethylene acetal 97%
3-Bromobenzaldehyde ethylene acetal
Formula:
C₉H₉BrO₂ MW: 229,07 g/mol Boiling Pt: 132…133 °C/8mmHg Density: 1,514 g/cm³ (25 °C) Flash Pt: >113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003209 CAS Number: 17789-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...