2-Bromoimidazo[1,2-a]pyridine
Supplier: Apollo Scientific
|
Warning
|
APOSOR301012-1GEA
84.4
EUR
APOSOR301012-1G
2-Bromoimidazo[1,2-a]pyridine
2-Bromoimidazo[1,2-a]pyridine
Formula:
C₇H₅BrN₂ MW: 197,0300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09834778 CAS Number: 112581-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...