2,4-Dichloro-5-methylpyridine 98%
|
Warning
|
Synonyms:
2,4-Dichloro-5-picoline
APOSOR301066-25GEA
0
EUR
APOSOR301066-25G
APOSOR301066-5G
2,4-Dichloro-5-methylpyridine 98%
2,4-Dichloro-5-methylpyridine
Formula:
C₆H₅Cl₂N MW: 162,02 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08704813 CAS Number: 56961-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...