Ethyl-2,4-dichloropyrimidine-5-carboxylate 98%

Supplier: Apollo Scientific
Warning

APOSOR301231-25GEA 81.4 EUR
APOSOR301231-25G APOSOR301231-100G
Ethyl-2,4-dichloropyrimidine-5-carboxylate 98%
Ethyl-2,4-dichloropyrimidine-5-carboxylate
Formula: C₇H₆Cl₂N₂O₂
MW: 221,04
Melting Pt: 36…37 °C
Storage Temperature: Ambient
MDL Number: MFCD09910281
CAS Number: 51940-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR