3-Chloro-2-methylbenzonitrile
|
Warning
|
APOSOR30171-5GEA
0
EUR
APOSOR30171-5G
APOSOR30171-25G
3-Chloro-2-methylbenzonitrile
3-Chloro-2-methylbenzonitrile
Formula:
C₈H₆ClN MW: 151,60 g/mol Boiling Pt: 120 °C/22mmHg Melting Pt: 20…23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045598 CAS Number: 54454-12-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...