3-(Benzyloxy)phenylboronic acid pinacol ester

Warning

Synonyms: 3-Benzyloxyphenylboronic acid, pinacol ester2-(3-(Benzyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane2-[3-(Benzyloxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

APOSOR303465-5GEA 0 EUR
APOSOR303465-5G APOSOR303465-500MG APOSOR303465-1G
3-(Benzyloxy)phenylboronic acid pinacol ester
3-(Benzyloxy)phenylboronic acid pinacol ester
Formula: C₁₉H₂₃BO₃
MW: 310,1951 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD05663839
CAS Number: 765908-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR