2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene

Synonyms: 9,9-Dioctylfluorene-2,7-diboronic acid bis(pinacol) ester2,2'-(9,9-Dioctyl-9H-fluorene-2,7-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

APOSOR303504-1GEA 0 EUR
APOSOR303504-1G APOSOR303504-500MG APOSOR303504-5G
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene
Formula: C₄₁H₆₄B₂O₄
MW: 642,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16294554
CAS Number: 196207-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR