4-(N,N-Diphenylamino)phenylboronic acid

Supplier: Apollo Scientific

Synonyms: 4-(Diphenylamino)benzeneboronic acid4-(Diphenylamino)phenylboronic acid

APOSOR303505-1GEA 125 EUR
APOSOR303505-1G APOSOR303505-5G APOSOR303505-500MG
4-(N,N-Diphenylamino)phenylboronic acid
4-(N,N-Diphenylamino)phenylboronic acid
Formula: C₁₈H₁₆BNO₂
MW: 289,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798117
CAS Number: 201802-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR