9-Anthraceneboronic acid

Supplier: Apollo Scientific

Synonyms: 9-anthraceneboronic acidanthracen-9-ylboronic acidAnthracene-9-boronic acid

APOSOR303566-1GEA 51 EUR
APOSOR303566-1G
9-Anthraceneboronic acid
9-Anthraceneboronic acid
Formula: C₁₄H₁₁BO₂
MW: 222,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03425925
CAS Number: 100622-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR