Ethyl isocyanatoacetate

Supplier: Apollo Scientific
Danger

APOSOR30501-5GEA 77.7 EUR
APOSOR30501-5G APOSOR30501-25G APOSOR30501-1G
Ethyl isocyanatoacetate
Ethyl isocyanatoacetate

  • May form small amount of sediment.

Formula: C₅H₇NO₃
MW: 129,12
Boiling Pt: 66…68 °C
Density: 1,151 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002040
CAS Number: 2949-22-6
UN: 2206
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR