4-Methyl-1H-indazole
APOSOR305100-250MGEA
58.2
EUR
APOSOR305100-250MG
APOSOR305100-1G
4-Methyl-1H-indazole
4-Methyl-1H-indazole
Formula:
C₈H₈N₂ MW: 132,1600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06200231 CAS Number: 3176-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...