4-tert-Butylphenylboronic acid pinacol ester 98%

Supplier: Apollo Scientific

Synonyms: 2-(4-(tert-Butyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

APOSOR30620-5GEA 68.4 EUR
APOSOR30620-5G
4-tert-Butylphenylboronic acid pinacol ester 98%
4-tert-Butylphenylboronic acid pinacol ester
Formula: C₁₆H₂₅BO₂
MW: 260,1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663843
CAS Number: 214360-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR