3-Amino-3-(3,4-dichlorophenyl)propionic acid

Supplier: Apollo Scientific

Synonyms: 3-Amino-3-(3,4-dichlorophenyl)propanoic acid

OR307537-1G
APOSOR307537-1GEA 259 EUR
APOSOR307537-1G
3-Amino-3-(3,4-dichlorophenyl)propionic acid
3-Amino-3-(3,4-dichlorophenyl)propionic acid
Formula: C₉H₉Cl₂NO₂
MW: 234,0793 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00462884
CAS Number: 117391-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR