7-Chloro-8-methylquinolin-4-ol

Supplier: Apollo Scientific

Synonyms: 7-Chloro-4-hydroxy-8-methylquinoline

APOSOR307830-5GEA 135 EUR
APOSOR307830-5G
7-Chloro-8-methylquinolin-4-ol
7-Chloro-8-methylquinolin-4-ol
Formula: C₁₀H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD00272440
CAS Number: 203626-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR