1-Methyl-1H-indazole-4-boronic acid pinacol ester

Synonyms: 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

APOSOR308053-1GEA 168 EUR
APOSOR308053-1G APOSOR308053-5G
1-Methyl-1H-indazole-4-boronic acid pinacol ester
1-Methyl-1H-indazole-4-boronic acid pinacol ester
Formula: C₁₄H₁₉BN₂O₂
MW: 258,1239 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10700158
CAS Number: 885698-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR