3-(4-Bromophenyl)propiononitrile 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(4-Bromophenyl)propanenitrile

APOSOR3087-5GEA 386 EUR
APOSOR3087-5G
3-(4-Bromophenyl)propiononitrile 98%
3-(4-Bromophenyl)propiononitrile
Formula: C₉H₈BrN
MW: 210,0705 g/mol
Boiling Pt: 118 °C/1mmHg
Density: 1,42 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01310795
CAS Number: 57775-08-3
UN: 3334
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR