2-Naphthyl phenyl ketone

Supplier: Apollo Scientific

Synonyms: 2-Benzoylnaphthalene

APOSOR310648-5GEA 157 EUR
APOSOR310648-5G
2-Naphthyl phenyl ketone
2-Naphthyl phenyl ketone
Formula: C₁₇H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00004106
CAS Number: 644-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR