N-(4-Methoxybenzylidene)-4-methoxyaniline
Synonyms:
N,1-bis(4-methoxyphenyl)methanimine
APOSOR311213-500MGEA
0
EUR
APOSOR311213-500MG
N-(4-Methoxybenzylidene)-4-methoxyaniline
N-(4-Methoxybenzylidene)-4-methoxyaniline
Formula:
C₁₅H₁₅NO₂ Melting Pt: 146…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025800 CAS Number: 1749-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...