1-Hydroxyethylferrocene

Supplier: Apollo Scientific
Warning

Synonyms: α-Hydroxyethylferrocene1-Ferrocenylethanol

APOSOR315064-10GEA 112 EUR
APOSOR315064-10G APOSOR315064-5G APOSOR315064-1G
1-Hydroxyethylferrocene
1-Hydroxyethylferrocene
Formula: C₁₂H₁₄FeO
MW: 230,09 g/mol
Melting Pt: 76…79 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00013752
CAS Number: 1277-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR